Order Entry
Denmark
Orders LinkContactUsLinkComponent
Overhead stirrer tools, anchor stirrers
Overhead stirrer tools, anchor stirrers
Anchor stirrer, DURAN®, 150 LF, 5000/10000 ml, 16 mm
Katalog # LENZ06321588
Leverandør:   Lenz Laborglas
Overhead stirrer tools, anchor stirrers
Katalog # LENZ06321588
Leverandør:  Lenz Laborglas
Overhead stirrer tools, anchor stirrers
Overhead stirrer tools, anchor stirrers
Anchor stirrer, DURAN®, 150 LF, 5000/10000 ml, 16 mm
Price
1.280,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Anchor stirrers with GL thread connection.
  • Made of precision calibrated glass tube
  • With ground and polished bearing surface
  • Suitable for flange vessels
Specifikationer
  • Description:
    Anchor stirrer, DURAN®, 150 LF
  • Shaft Ø:
    16 mm
  • For container:
    5000/10000 ml
Anbefalinger