Order Entry
Denmark
Orders LinkContactUsLinkComponent
Overhead stirrer tools, anchor stirrers

Overhead stirrer tools, anchor stirrers

Anchor stirrer, DURAN®, 100 LF, 3000 ml, 10 mm
Katalog # LENZ06321076
Leverandør:   Lenz Laborglas
Overhead stirrer tools, anchor stirrers
Katalog # LENZ06321076
Leverandør:  Lenz Laborglas
Overhead stirrer tools, anchor stirrers
Overhead stirrer tools, anchor stirrers

Anchor stirrer, DURAN®, 100 LF, 3000 ml, 10 mm

Price
kr1,070.00
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Anchor stirrers with GL thread connection.

  • Made of precision calibrated glass tube
  • With ground and polished bearing surface
  • Suitable for flange vessels

Documents

Specifikationer
  • Description:
    Anchor stirrer, DURAN®, 100 LF
  • Shaft Ø:
    10 mm
  • For container:
    3000 ml
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.