Order Entry
Denmark
Orders LinkContactUsLinkComponent
Overhead stirrer tools, anchor stirrers

Overhead stirrer tools, anchor stirrers

Anchor stirrer, DURAN®, 60 LF, 100 ml, 10 mm
Catalog # LENZ06320637
Supplier:   Lenz Laborglas
Overhead stirrer tools, anchor stirrers
Catalog # LENZ06320637
Supplier:  Lenz Laborglas
Overhead stirrer tools, anchor stirrers
Overhead stirrer tools, anchor stirrers

Anchor stirrer, DURAN®, 60 LF, 100 ml, 10 mm

Price
1.020,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Anchor stirrers with GL thread connection.

  • Made of precision calibrated glass tube
  • With ground and polished bearing surface
  • Suitable for flange vessels

Documents

Specifications
  • Description:
    Anchor stirrer, DURAN®, 60 LF
  • Shaft Ø:
    10 mm
  • For container:
    100 ml