Raschig rings
Raschig ring, 5 mm, 5 mm
Catalog # LENZ05127005
Supplier: Lenz Laborglas
Jump to:
Product Details & Documents
Soda lime glass.
- For packing columns
- Good chemical resistance
- Good thermal resistance
Specifications
- Ext.Ø:5 mm
- Description:Raschig ring
- Height:5 mm