Order Entry
Denmark
Orders LinkContactUsLinkComponent
Chromatography columns, Flex-Column®, KIMBLE®

Chromatography columns, Flex-Column®, KIMBLE®

Chromatography column, 25,00 mm, 100 mm, 49 ml
Catalog # KIML420400-2510
Chromatography columns, Flex-Column®, KIMBLE®
Catalog # KIML420400-2510
Supplier:  
Chromatography columns, Flex-Column®, KIMBLE®
Chromatography columns, Flex-Column®, KIMBLE®

Chromatography column, 25,00 mm, 100 mm, 49 ml

Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
FLEX-COLUMN ECONOMY 2.5X10CM TEMP 0-50°C 1 * 5 items Catalog #SCERSP420400-2510

Product SCERSP420400-2510 is a replacement for KIML420400-2510 but varies by Brand, Packaging or Quantity.

Product Details & Documents

These economical columns are constructed with borosilicate glass barrels, molded PP end caps, and PE bed supports. They also feature Luer lock inlet and outlet fittings for better connections.

PP packing reservoir funnels are for use in initial column packing or as small buffer reservoirs. Glass packing reservoirs are ideal for packing gel slurries into columns or acting as a buffer reservoir in gravity flow chromatography. Glass reservoirs include a PP upper cap with male Luer lock fitting.

Flow adapters allow easy adjustment of column bed volume and offer improved column performance and chromatogram resolution. Flow adapters are constructed with a PTFE eluant cannula, a Delrin® body, a Viton® O-ring seal, and a 20 µm PE bed support. Each flow adapter includes one replacment bed support; additional replacements are also available.

Specifications
  • Ø int.:
    25,00 mm
  • Capacity:
    49 ml
  • Description:
    Chromatography column
  • Length:
    100 mm
  • Seq:
    08
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.