Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Double spatulas, micro, Chattaway type

Stainless steel, 180 mm, 4 mm
  NICK2353
Double spatulas, micro, Chattaway type
  NICK2353
Double spatulas, micro, Chattaway type
Double spatulas, micro, Chattaway type

Stainless steel, 180 mm, 4 mm

 
kr53.40
   
1 * 1 Items
 

Stainless steel or pure nickel (99,6%). Multipurpose spatula for a wide range of uses.

  • One end is flat and spatula-shaped, the other end is rounded and turned up at 45°
  • Excellent chemical resistance
  • Excellent thermal resistance
  • Quick and easy to clean
 
  • Description:
    Stainless steel
  • Length:
    180 mm
  • Blade width:
    4 mm
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.