Order Entry
Denmark
Orders LinkContactUsLinkComponent
Adapters, NS cone to spherical joint socket
Adapters, NS cone to spherical joint socket
29/32 NS, S 19
Catalog # LENZ01521929
Supplier:   Lenz Laborglas
Adapters, NS cone to spherical joint socket
Catalog # LENZ01521929
Supplier:  Lenz Laborglas
Adapters, NS cone to spherical joint socket
Adapters, NS cone to spherical joint socket
29/32 NS, S 19
Price
327,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
DURAN®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Adapters from ground joint cone to spherical joint socket.
Documents
Specifications
  • Cone size:
    29/32 NS
  • Socket:
    S 19
  • Seq:
    02