Order Entry
Denmark
Orders LinkContactUsLinkComponent
Magnetic stirring bars, oval

Magnetic stirring bars, oval

40 mm, 13 mm
Katalog # COWI001.640.13
Leverandør:   COWIE
Magnetic stirring bars, oval
Katalog # COWI001.640.13
Leverandør:  COWIE
Magnetic stirring bars, oval
Magnetic stirring bars, oval

40 mm, 13 mm

Price
kr48.00
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

PTFE covered. For stirrers with rare earth magnetic core the magnetic attraction is 1,5 to 2 times stronger than conventional stirrers. The magnetic core is made from a combination of samarium and cobalt. The rare earth stirrers will continue to function at full strength for many years as the magnetism remains for longer.

  • Very good chemical resistance
  • Temperature resistant, suitable for use at high and low temperatures (−200 to +280 °C)
Specifikationer
  • Ø:
    13 mm
  • Length:
    40 mm
  • Seq:
    015
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.