Order Entry
Denmark
Orders LinkContactUsLinkComponent
Clamps
Flat flanges DN 150
  LENZ06247015
Clamps
  LENZ06247015
Clamps
Clamps
Flat flanges DN 150
14.070,00 kr
1 * 1 Items
Stainless steel.
  • Excellent chemical resistance
  • Excellent temperature resistance
For fixing lids onto reaction vessels, available for flat flanges LF100-200.
  • For:
    Flat flanges DN 150