1,1-Dimethoxy-2-phenylethan ≥98%
Glass bottle for solids, 500 g
Supplier: Thermo Fisher Scientific
1,1-Dimethoxy-2-phenylethan ≥98%
Catalog # A18730.36
Supplier: Thermo Fisher Scientific
CAS Number: 101-48-4
Jump to:
Product Details & Documents
Purity: 98% CAS No.: 101-48-4 Molecular Formula: C10H14O2 Formula Weight: 166.22 Flash Point: 83°(181°F) Refractive Index: 1.4940 Boiling Point: 220° Beilstein Reference: 879360 MDL No.: MFCD00008487
Synonyms: Phenylacetaldehyde dimethyl acetal
- CAS Number:101-48-4
- Formula:C₆H₅CH₂CH(OCH₃)₂
- MDL Number:MFCD00008487
- Molecular Weight:166.22 g/mol
- Boiling Pt:220 °C
- Melting Pt:62…64 °C
- Density:1,004
- Flash Pt:83 °C (181 °F)
- Storage Temperature:Stue temperatur
- EINECS:202-945-6
Specifications
Specifications
- Pakning:Glass bottle for solids
- Pakn:500 g



