Order Entry
Denmark
Orders LinkContactUsLinkComponent
1,1-Dimethoxy-2-phenylethane ≥98%

1,1-Dimethoxy-2-phenylethane ≥98%

Glass bottle for solids, 100 g
Catalog # A18730.22
1,1-Dimethoxy-2-phenylethane ≥98%
Catalog # A18730.22
CAS Number:  101-48-4
1,1-Dimethoxy-2-phenylethane ≥98%
1,1-Dimethoxy-2-phenylethane ≥98%

Glass bottle for solids, 100 g

CAS Number:  101-48-4
Price
359,00 kr
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 98% CAS No.: 101-48-4 Molecular Formula: C10H14O2 Formula Weight: 166.22 Flash Point: 83°(181°F) Refractive Index: 1.4940 Boiling Point: 220° Beilstein Reference: 879360 MDL No.: MFCD00008487

Synonyms:  Phenylacetaldehyde dimethyl acetal
  • CAS Number:101-48-4
  • Formula:C₆H₅CH₂CH(OCH₃)₂
  • MDL Number:MFCD00008487
  • Molecular Weight:166.22 g/mol
  • Boiling Pt:220 °C
  • Melting Pt:62…64 °C
  • Density:1.004
  • Flash Pt:83 °C (181 °F)
  • Storage Temperature:Ambient
  • EINECS:202-945-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.