Order Entry
Denmark
Orders LinkContactUsLinkComponent
Bromophenol blue

Bromophenol blue

Glass bottle for solids, 10 g
Catalog # A18469.09
Bromophenol blue
Catalog # A18469.09
CAS Number:  115-39-9
Bromophenol blue
Bromophenol blue

Glass bottle for solids, 10 g

CAS Number:  115-39-9
Price
541,00 kr
Log in for availability
1 * 10 g
Product Details & Documents

CAS No.: 115-39-9 Molecular Formula: C19H10Br4O5S Formula Weight: 669.98 Odour: Odourless Melting Point: ca 270° dec. Beilstein Reference: 61698 Merck Reference: 14,1444 MDL No.: MFCD00005875

Synonyms:  Bromphenol Blue Sultone Form, BPB, 3,3',5,5'-Tetrabromophenolsulphonphthalein, Tetrabromophenol-sulphonephthalein
  • CAS Number:115-39-9
  • Formula:C₁₉H₁₀Br₄O₅S
  • MDL Number:MFCD00005875
  • Molecular Weight:669.97 g/mol
  • Merck Index:14,1444
  • Melting Pt:∼ 270 °C (dec.)
  • Density:1 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:204-086-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications