Order Entry
Denmark
Orders LinkContactUsLinkComponent
Galactitol ≥97%

Galactitol ≥97%

Glass bottle for solids, 25 g
Catalog # A18402.14
Galactitol ≥97%
Catalog # A18402.14
CAS Number:  608-66-2
Galactitol ≥97%
Galactitol ≥97%

Glass bottle for solids, 25 g

CAS Number:  608-66-2
Price
309,00 kr
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 97% CAS No.: 608-66-2 Molecular Formula: C6H14O6 Formula Weight: 182.17 Melting Point: 187-191° Boiling Point: 275-280°/1mm Merck Reference: 14,4332 MDL No.: MFCD00064288

Synonyms:  Dulcitol, rel-(2R,3S,4R,5S)-Hexane-1,2,3,4,5,6-hexaol
  • CAS Number:608-66-2
  • Formula:HOH₂C(CH(OH))₄CH₂OH
  • MDL Number:MFCD00064288
  • Molecular Weight:182.17 g/mol
  • Merck Index:14,4332
  • Boiling Pt:275…280 °C (1 mmHg)
  • Melting Pt:187…191 °C
  • Density:1.47
  • Storage Temperature:Ambient
  • EINECS:210-165-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.