Murexide
Glass bottle for solids, 50 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 3051-09-0 Molecular Formula: C8H8N6O6 Formula Weight: 284.19 Melting Point: 300° Beilstein Reference: 3582175 Merck Reference: 14,6306 MDL No.: MFCD00012777
Synonyms: Ammonium purpurate, Murexide triturate, Ammonium 5-(2,4,6-trioxoperhydropyrimidin-5-ylideneamino)barbiturate, Purpuric acid ammonium salt
- CAS Number:3051-09-0
- Formula:C₈H₈N₆O₆
- MDL Number:MFCD00012777
- Molecular Weight:284.19 g/mol
- Merck Index:14,6306
- Melting Pt:>300 °C
- Storage Temperature:Ambient
- EINECS:221-266-6
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g



