Order Entry
Denmark
Orders LinkContactUsLinkComponent
Acrylamide ≥98%

Acrylamide ≥98%

Glass bottle for solids, 500 g
Catalog # A17157.36
Toxic
Health
Irritant
Acrylamide ≥98%
Catalog # A17157.36
CAS Number:  79-06-1
Acrylamide ≥98%
Acrylamide ≥98%

Glass bottle for solids, 500 g

CAS Number:  79-06-1
Price
507,00 kr
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 98+% CAS No.: 79-06-1 Molecular Formula: CH2=CHCONH2 Formula Weight: 71.08 Flash Point: 138°(280°F) Odour: Odourless Boiling Point: 125°/25mm Beilstein Reference: 605349 Merck Reference: 14,129 UN No.: 2074 MDL No.: MFCD00008032

Synonyms:  Prop-2-enamide
  • CAS Number:79-06-1
  • Formula:C₃H₅NO
  • MDL Number:MFCD00008032
  • Molecular Weight:71.08 g/mol
  • Merck Index:14,129
  • Boiling Pt:125 °C (25 mmHg)
  • Melting Pt:82…85 °C
  • Density:1.322
  • Flash Pt:138 °C (280 °F)
  • Storage Temperature:Ambient
  • EINECS:201-173-7
  • UN:2074
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.