Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Valeric acid ≥99%

Glass bottle, 100 mL
Catalog # A16238.AE
Corrosive
Irritant
Valeric acid ≥99%
Catalog # A16238.AE
CAS Number:  109-52-4
Valeric acid ≥99%
Valeric acid ≥99%

Glass bottle, 100 mL

CAS Number:  109-52-4
Price
362,00 kr
Log in for availability
1 * 100 mL
Product Details & Documents

Purity: 99% CAS No.: 109-52-4 Molecular Formula: C5H10O2 Formula Weight: 102.13 Flash Point: 88°(190°F) Refractive Index: 1.4080 Odour: Unpleasant Melting Point: -34° Boiling Point: 184-186° Beilstein Reference: 969454 Merck Reference: 14,9904 UN No.: 3265 MDL No.: MFCD00004413

Synonyms:  Pentanoic acid
  • CAS Number:109-52-4
  • Formula:C₅H₁₀O₂
  • MDL Number:MFCD00004413
  • Molecular Weight:102.13 g/mol
  • Merck Index:14,9904
  • Boiling Pt:184…186 °C
  • Melting Pt:–34 °C
  • Density:0.938
  • Flash Pt:88 °C (190 °F)
  • Storage Temperature:Ambient
  • EINECS:203-677-2
  • UN:3265
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.