Valeric acid ≥99%
Glass bottle, 2.500 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 109-52-4 Molecular Formula: C5H10O2 Formula Weight: 102.13 Flash Point: 88°(190°F) Refractive Index: 1.4080 Odour: Unpleasant Melting Point: -34° Boiling Point: 184-186° Beilstein Reference: 969454 Merck Reference: 14,9904 UN No.: 3265 MDL No.: MFCD00004413
Synonyms: Pentanoic acid
- CAS Number:109-52-4
- Formula:C₅H₁₀O₂
- MDL Number:MFCD00004413
- Molecular Weight:102.13 g/mol
- Merck Index:14,9904
- Boiling Pt:184…186 °C
- Melting Pt:–34 °C
- Density:0.938
- Flash Pt:88 °C (190 °F)
- Storage Temperature:Ambient
- EINECS:203-677-2
- UN:3265
- ADR:8,III
Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:2.500 mL
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



