Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

1-Octanol ≥99%

Glass bottle, 2.500 mL
Catalog # A15977.0F
Irritant
1-Octanol ≥99%
Catalog # A15977.0F
CAS Number:  111-87-5
1-Octanol ≥99%
1-Octanol ≥99%

Glass bottle, 2.500 mL

CAS Number:  111-87-5
Price
1.290,00 kr
Log in for availability
1 * 2.500 mL
Product Details & Documents

Purity: 99% CAS No.: 111-87-5 Molecular Formula: C8H18O Formula Weight: 130.23 Flash Point: 81°(177°F) Physical Form: Liquid Density: 0.825 Refractive Index: 1.4290 Appearance: Colourless Odour: Sweet Melting Point: -16° Boiling Point: 193-195° Beilstein Reference: 1697461 Merck Reference: 14,6751 MDL No.: MFCD00002988

Synonyms:  n-Octanol
  • CAS Number:111-87-5
  • Formula:H₃C(CH₂)₆CH₂OH
  • MDL Number:MFCD00002988
  • Molecular Weight:130.23 g/mol
  • Merck Index:14,6751
  • Boiling Pt:193…195 °C
  • Melting Pt:–16 °C
  • Density:0.825
  • Flash Pt:81 °C (177 °F)
  • Storage Temperature:Ambient
  • EINECS:203-917-6
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2.500 mL

Specifications