Eugenol ≥99%
Glass bottle for solids, 500 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 97-53-0 Molecular Formula: C10H12O2 Formula Weight: 164.21 Flash Point: 127°(260°F) Refractive Index: 1.5410 Odour: Pungent Melting Point: -10° Boiling Point: 252-253° Beilstein Reference: 1366759 Merck Reference: 14,3898 MDL No.: MFCD00008654
Synonyms: 4-Allyl-2-methoxyphenol, 5-Allyl-2-hydroxyanisole
- CAS Number:97-53-0
- Formula:C₁₀H₁₂O₂
- MDL Number:MFCD00008654
- Molecular Weight:164.2 g/mol
- Merck Index:14,3898
- Boiling Pt:252…253 °C
- Melting Pt:–10 °C
- Density:1.068
- Flash Pt:127 °C (260 °F)
- Storage Temperature:Ambient
- EINECS:202-589-1
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



