Order Entry
Denmark
Orders LinkContactUsLinkComponent
D-(-)-Mannitol ≥97.0%, white powder

D-(-)-Mannitol ≥97.0%, white powder

Glass bottle for solids, 1.000 g
Catalog # A14030.0B
D-(-)-Mannitol ≥97.0%, white powder
Catalog # A14030.0B
CAS Number:  69-65-8
D-(-)-Mannitol ≥97.0%, white powder
D-(-)-Mannitol ≥97.0%, white powder

Glass bottle for solids, 1.000 g

CAS Number:  69-65-8
Price
931,00 kr
Log in for availability
1 * 1.000 g
Product Details & Documents

Purity: 99% CAS No.: 69-65-8 Molecular Formula: C6H14O6 Formula Weight: 182.17 Odour: Odourless Melting Point: 165-167° Boiling Point: 290-295°/3.5mm Beilstein Reference: 1721898 Merck Reference: 14,5745 Optical Rotation: +24° (in borax solution) MDL No.: MFCD00064287

Synonyms:  Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol
  • CAS Number:69-65-8
  • Formula:HOH₂C(CH(OH))₄CH₂OH
  • MDL Number:MFCD00064287
  • Molecular Weight:182.17 g/mol
  • Merck Index:14,5745
  • Boiling Pt:290…295 °C (3.5 mmHg)
  • Melting Pt:165…167 °C
  • Density:1.489
  • Storage Temperature:Ambient
  • EINECS:200-711-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1.000 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.