Order Entry
Denmark
Orders LinkContactUsLinkComponent
Pyrogallol ≥98%
Glass bottle for solids, 500 g
  A13405.36
Pyrogallol ≥98%
  A13405.36
 :  87-66-1
Pyrogallol ≥98%
Pyrogallol ≥98%
Glass bottle for solids, 500 g
 :  87-66-1
2.400,00 kr
1 * 500 g
Purity: 98+% CAS No.: 87-66-1 Molecular Formula: C6H6O3 Formula Weight: 126.11 Odour: Odourless Melting Point: 131-135° Boiling Point: 309° Beilstein Reference: 907431 Merck Reference: 14,8000 UN No.: 2811 MDL No.: MFCD00002192
 :  Benzene-1,2,3-triol, 1,2,3-Trihydroxybenzene
  •  :87-66-1
  •  :C₆H₆O₃
  •  :MFCD00002192
  •  :126.11 g/mol
  •  :14,8000
  •  :309 °C
  •  :131…135 °C
  •  :1.45
  •  :Ambient
  •  :201-762-9
  •  :2811
  •  :6.1,III
  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g