Order Entry
Denmark
Orders LinkContactUsLinkComponent
Pyrogallol ≥98%

Pyrogallol ≥98%

Glass bottle for solids, 100 g
Katalog # A13405.22
Health
Irritant
Leverandør:   Thermo Fisher Scientific
Pyrogallol ≥98%
Katalog # A13405.22
CAS nummer:  87-66-1
Pyrogallol ≥98%
Pyrogallol ≥98%

Glass bottle for solids, 100 g

CAS nummer:  87-66-1
Price
722,00 kr
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 98+% CAS No.: 87-66-1 Molecular Formula: C6H6O3 Formula Weight: 126.11 Odour: Odourless Melting Point: 131-135° Boiling Point: 309° Beilstein Reference: 907431 Merck Reference: 14,8000 UN No.: 2811 MDL No.: MFCD00002192

Synonyms:  Benzene-1,2,3-triol, 1,2,3-Trihydroxybenzene
  • CAS Number:87-66-1
  • Formula:C₆H₆O₃
  • MDL Number:MFCD00002192
  • Molecular Weight:126.11 g/mol
  • Merck Index:14,8000
  • Boiling Pt:309 °C
  • Melting Pt:131…135 °C
  • Density:1.45
  • Storage Temperature:Ambient
  • EINECS:201-762-9
  • UN:2811
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications