Order Entry
Denmark
Orders LinkContactUsLinkComponent
Resorcinol ≥99%

Resorcinol ≥99%

Glass bottle for solids, 50 g
Catalog # A13080.18
Irritant
Environment
Resorcinol ≥99%
Catalog # A13080.18
CAS Number:  108-46-3
Resorcinol ≥99%
Resorcinol ≥99%

Glass bottle for solids, 50 g

CAS Number:  108-46-3
Price
377,00 kr
Log in for availability
1 * 50 g
Product Details & Documents

Purity: 99% CAS No.: 108-46-3 Molecular Formula: C6H6O2 Formula Weight: 110.11 Flash Point: 171°(339°F) Odour: Phenol-like pH: 4-6 Melting Point: 109-112° Boiling Point: 280-281° Beilstein Reference: 906905 Merck Reference: 14,8155 UN No.: 2876 MDL No.: MFCD00002269

Synonyms:  1,3-Dihydroxybenzene, 1,3-Benzenediol
  • CAS Number:108-46-3
  • Formula:C₆H₆O₂
  • MDL Number:MFCD00002269
  • Molecular Weight:110.11 g/mol
  • Merck Index:14,8155
  • Boiling Pt:280…281 °C
  • Melting Pt:109…112 °C
  • Density:1.272
  • Flash Pt:171 °C (339 °F)
  • Storage Temperature:Ambient
  • EINECS:203-585-2
  • UN:2876
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.