Sodium fluoride ≥99%
Glass bottle for solids, 1.000 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 7681-49-4 Molecular Formula: FNa Formula Weight: 41.99 Refractive Index: 1.336 Odour: Odourless Boiling Point: 1704° Merck Reference: 14,8618 UN No.: 1690 MDL No.: MFCD00003524
- CAS Number:7681-49-4
- Formula:NaF
- MDL Number:MFCD00003524
- Molecular Weight:41.99 g/mol
- Merck Index:14,8618
- Boiling Pt:1704 °C
- Melting Pt:993 °C
- Density:2.78
- Storage Temperature:Ambient
- EINECS:231-667-8
- UN:1690
- ADR:6.1,III
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1.000 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



