Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Fuchsin (acid)

Glass bottle for solids, 100 g
Catalog # B22222.22
Irritant
Fuchsin (acid)
Catalog # B22222.22
CAS Number:  3244-88-0
Fuchsin (acid)
Fuchsin (acid)

Glass bottle for solids, 100 g

CAS Number:  3244-88-0
Price
2.930,00 kr
Log in for availability
1 * 100 g
Product Details & Documents

CAS No.: 3244-88-0 Molecular Formula: C20H17N3Na2O9S3 Formula Weight: 585.53 Beilstein Reference: 4111656 Merck Reference: 14,107 MDL No.: MFCD00013286

Synonyms:  Rubin S, CN DcR-8, Acid Violet 19, Acid Fuchsin, Acid Fuchsin sodium salt, Fuchsin S, CI 42685, Acid Magenta, 3-(1-(4-Amino-3-methyl-5-sulphonatophenyl)-1-(4-amino-3-sulphonatophenyl)methylene)cyclohexa-1,4-dienesulphonic acid, Benzenesulphonic acid, 2-amino-5-[(4-amino-3-sulphophenyl)(4-imino-3-sulpho-2,5-cyclohexadien-1-ylidene)methyl]-3-methyl-, sodium salt (1:2)
  • CAS Number:3244-88-0
  • Formula:C₂₀H₁₇N₃Na₂O₉S₃
  • MDL Number:MFCD00013286
  • Molecular Weight:585.55 g/mol
  • Merck Index:14,107
  • Melting Pt:> 130 °C
  • Storage Temperature:Ambient
  • EINECS:221-816-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications