Squalene ≥98%
Glass bottle for solids, 500 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 98% CAS No.: 111-02-4 Molecular Formula: C30H50 Formula Weight: 410.73 Storage Temperature: 2°C to 8°C Flash Point: 218°(424°F) Refractive Index: 1.4955 Melting Point: -75° Boiling Point: 285°/25mm Merck Reference: 14,8768 MDL No.: MFCD00008912
Biosynthetic precursor to all steroids
Synonyms: 2,6,10,15,19,23-Hexamethyltetracosa-2,6,10,14,18,22-hexaene
- CAS Number:111-02-4
- Formula:C₃₀H₅₀
- MDL Number:MFCD00008912
- Molecular Weight:410.73 g/mol
- Merck Index:14,8768
- Boiling Pt:285 °C (25 mmHg)
- Melting Pt:–75 °C
- Density:0.855
- Flash Pt:218 °C (424 °F)
- EINECS:203-826-1
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



