Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Thiourea ≥99%

Glass bottle for solids, 10.000 g
Catalog # A12828.0C
Health
Irritant
Environment
Thiourea ≥99%
Catalog # A12828.0C
CAS Number:  62-56-6
Thiourea ≥99%
Thiourea ≥99%

Glass bottle for solids, 10.000 g

CAS Number:  62-56-6
Price
5.720,00 kr
Log in for availability
1 * 10.000 g
Product Details & Documents

Purity: 99% CAS No.: 62-56-6 Molecular Formula: CH4N2S Formula Weight: 76.12 Odour: Odourless Melting Point: 172-178° Beilstein Reference: 605327 Merck Reference: 14,9367 UN No.: 3077 MDL No.: MFCD00008067

Synonyms:  Thiocarbamide, Sulphourea
  • CAS Number:62-56-6
  • Formula:NH₂CSNH₂
  • MDL Number:MFCD00008067
  • Molecular Weight:76.12 g/mol
  • Merck Index:14,9367
  • Melting Pt:172…178 °C
  • Density:1.405
  • Storage Temperature:Ambient
  • EINECS:200-543-5
  • UN:3077
  • ADR:9,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10.000 g

Specifications