Order Entry
Denmark
Orders LinkContactUsLinkComponent
2-Thiobarbituric acid ≥98%
Glass bottle for solids, 500 g
  A12681.36
2-Thiobarbituric acid ≥98%
  A12681.36
 :  504-17-6
2-Thiobarbituric acid ≥98%
2-Thiobarbituric acid ≥98%
Glass bottle for solids, 500 g
 :  504-17-6
4.900,00 kr
1 * 500 g
Purity: 98% CAS No.: 504-17-6 Molecular Formula: C4H4N2O2S Formula Weight: 144.15 pH: 1.8 Melting Point: 300° Beilstein Reference: 120663 MDL No.: MFCD00006674
 :  4,6-Dihydroxy-2-mercaptopyrimidine
  •  :504-17-6
  •  :C₄H₄N₂O₂S
  •  :MFCD00006674
  •  :144.15 g/mol
  •  :419 °C (1013 hPa)
  •  :235 °C
  •  :1.661 g/cm³ (20 °C)
  •  :Ambient
  •  :207-985-8
  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g