Order Entry
Denmark
Orders LinkContactUsLinkComponent
2-Thiobarbituric acid ≥98%

2-Thiobarbituric acid ≥98%

Glass bottle for solids, 100 g
Catalog # A12681.22
2-Thiobarbituric acid ≥98%
Catalog # A12681.22
CAS Number:  504-17-6
2-Thiobarbituric acid ≥98%
2-Thiobarbituric acid ≥98%

Glass bottle for solids, 100 g

CAS Number:  504-17-6
Price
1.500,00 kr
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 98% CAS No.: 504-17-6 Molecular Formula: C4H4N2O2S Formula Weight: 144.15 pH: 1.8 Melting Point: 300° Beilstein Reference: 120663 MDL No.: MFCD00006674

Synonyms:  4,6-Dihydroxy-2-mercaptopyrimidine
  • CAS Number:504-17-6
  • Formula:C₄H₄N₂O₂S
  • MDL Number:MFCD00006674
  • Molecular Weight:144.15 g/mol
  • Boiling Pt:419 °C (1013 hPa)
  • Melting Pt:235 °C
  • Density:1.661 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:207-985-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications