Order Entry
Denmark
Orders LinkContactUsLinkComponent
(S)-(-)-Nicotine ≥99%

(S)-(-)-Nicotine ≥99%

Glass bottle for solids, 500 g
Catalog # A12398.36
Toxic
Environment
(S)-(-)-Nicotine ≥99%
Catalog # A12398.36
CAS Number:  54-11-5
(S)-(-)-Nicotine ≥99%
(S)-(-)-Nicotine ≥99%

Glass bottle for solids, 500 g

CAS Number:  54-11-5
Price
8.860,00 kr
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 99% CAS No.: 54-11-5 Molecular Formula: C10H14N2 Formula Weight: 162.24 Flash Point: 101°(213°F) Refractive Index: 1.5265 Odour: Characteristic pH: 10.2 Melting Point: -79° Boiling Point: 245-248° Beilstein Reference: 82109 Merck Reference: 14,6524 Optical Rotation: -168° (neat) UN No.: 1654 MDL No.: MFCD00006369

Synonyms:  (S)-(-)-3-(N-Methyl-2-pyrrolidinyl)pyridine, (S)-(-)-3-(1-Methyl-2-pyrrolidinyl)pyridine, L(-)-Nicotine, L-Nicotine, Nicotine, (-)-Nicotine
  • CAS Number:54-11-5
  • Formula:C₁₀H₁₄N₂
  • MDL Number:MFCD00006369
  • Molecular Weight:162.23 g/mol
  • Merck Index:14,6524
  • Boiling Pt:245…248 °C
  • Melting Pt:–79 °C
  • Density:1.017
  • Flash Pt:101 °C (213 °F)
  • Storage Temperature:Ambient
  • EINECS:200-193-3
  • UN:1654
  • ADR:6.1,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.