Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

(S)-(-)-Nicotine ≥99%

Glass bottle for solids, 100 g
Catalog # A12398.22
Toxic
Environment
(S)-(-)-Nicotine ≥99%
Catalog # A12398.22
CAS Number:  54-11-5
(S)-(-)-Nicotine ≥99%
(S)-(-)-Nicotine ≥99%

Glass bottle for solids, 100 g

CAS Number:  54-11-5
Price
1.920,00 kr
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 99% CAS No.: 54-11-5 Molecular Formula: C10H14N2 Formula Weight: 162.24 Flash Point: 101°(213°F) Refractive Index: 1.5265 Odour: Characteristic pH: 10.2 Melting Point: -79° Boiling Point: 245-248° Beilstein Reference: 82109 Merck Reference: 14,6524 Optical Rotation: -168° (neat) UN No.: 1654 MDL No.: MFCD00006369

Synonyms:  (S)-(-)-3-(N-Methyl-2-pyrrolidinyl)pyridine, (S)-(-)-3-(1-Methyl-2-pyrrolidinyl)pyridine, L(-)-Nicotine, L-Nicotine, Nicotine, (-)-Nicotine
  • CAS Number:54-11-5
  • Formula:C₁₀H₁₄N₂
  • MDL Number:MFCD00006369
  • Molecular Weight:162.23 g/mol
  • Merck Index:14,6524
  • Boiling Pt:245…248 °C
  • Melting Pt:–79 °C
  • Density:1.017
  • Flash Pt:101 °C (213 °F)
  • Storage Temperature:Ambient
  • EINECS:200-193-3
  • UN:1654
  • ADR:6.1,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications