Order Entry
Denmark
Orders LinkContactUsLinkComponent
(S)-(-)-Nicotine ≥99%
(S)-(-)-Nicotine ≥99%
Glass bottle for solids, 25 g
  A12398.14
(S)-(-)-Nicotine ≥99%
  A12398.14
 :  54-11-5
(S)-(-)-Nicotine ≥99%
(S)-(-)-Nicotine ≥99%
Glass bottle for solids, 25 g
 :  54-11-5
718,00 kr
1 * 25 g
Purity: 99% CAS No.: 54-11-5 Molecular Formula: C10H14N2 Formula Weight: 162.24 Flash Point: 101°(213°F) Refractive Index: 1.5265 Odour: Characteristic pH: 10.2 Melting Point: -79° Boiling Point: 245-248° Beilstein Reference: 82109 Merck Reference: 14,6524 Optical Rotation: -168° (neat) UN No.: 1654 MDL No.: MFCD00006369
 :  (S)-(-)-3-(N-Methyl-2-pyrrolidinyl)pyridine, (S)-(-)-3-(1-Methyl-2-pyrrolidinyl)pyridine, L(-)-Nicotine, L-Nicotine, Nicotine, (-)-Nicotine
  •  :54-11-5
  •  :C₁₀H₁₄N₂
  •  :MFCD00006369
  •  :162.23 g/mol
  •  :14,6524
  •  :245…248 °C
  •  :–79 °C
  •  :1.017
  •  :101 °C (213 °F)
  •  :Ambient
  •  :200-193-3
  •  :1654
  •  :6.1,II
  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g