Stearic acid ≥98%
Glass bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 98% CAS No.: 57-11-4 Molecular Formula: CH3(CH2)16CO2H Formula Weight: 284.48 Flash Point: 196°(384°F) Physical Form: Flakes or powder Density: 0.941 Appearance: White Odour: Odourless Melting Point: 69-72° Boiling Point: 232°/15mm Beilstein Reference: 608585 Merck Reference: 14,8804 MDL No.: MFCD00002752
A saturated fatty acid
Synonyms: Octadecanoic acid, Octadecylic acid
- CAS Number:57-11-4
- Formula:C₁₈H₃₆O₂
- MDL Number:MFCD00002752
- Molecular Weight:284.48 g/mol
- Merck Index:14,8804
- Boiling Pt:232 °C (15 mmHg)
- Melting Pt:69…72 °C
- Density:0.941
- Flash Pt:196 °C (384 °F)
- Storage Temperature:Ambient
- EINECS:200-313-4
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



