Order Entry
Denmark
Orders LinkContactUsLinkComponent
Vanillic acid ≥98%

Vanillic acid ≥98%

Glass bottle for solids, 100 g
Catalog # A12074.22
Irritant
Vanillic acid ≥98%
Catalog # A12074.22
CAS Number:  121-34-6
Vanillic acid ≥98%
Vanillic acid ≥98%

Glass bottle for solids, 100 g

CAS Number:  121-34-6
Price
1.390,00 kr
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 98% CAS No.: 121-34-6 Molecular Formula: C8H8O4 Formula Weight: 168.15 Melting Point: 210-218° Beilstein Reference: 2208364 Merck Reference: 14,9931 MDL No.: MFCD00002551

Synonyms:  4-Hydroxy-3-methoxybenzoic acid
  • CAS Number:121-34-6
  • Formula:C₈H₈O₄
  • MDL Number:MFCD00002551
  • Molecular Weight:168.15 g/mol
  • Merck Index:14,9931
  • Boiling Pt:353 °C (1013 hPa)
  • Melting Pt:210…218 °C
  • Storage Temperature:Ambient
  • EINECS:204-466-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications