Order Entry
Denmark
Orders LinkContactUsLinkComponent
Myristic acid ≥98%

Myristic acid ≥98%

Glass bottle for solids, 250 g
Katalog # A12067.30
Irriterende
Leverandør:   Thermo Fisher Scientific
Myristic acid ≥98%
Katalog # A12067.30
CAS nummer:  544-63-8
Myristic acid ≥98%
Myristic acid ≥98%

Glass bottle for solids, 250 g

CAS nummer:  544-63-8
Price
271,00 kr
Log ind for tilgængelighed
1 * 250 g
Product Details & Documents

Purity: 98% CAS No.: 544-63-8 Molecular Formula: CH3(CH2)12CO2H Formula Weight: 228.38 Flash Point: 175°(347°F) Melting Point: 53-56° Boiling Point: 326° Beilstein Reference: 508624 Merck Reference: 14,6333 MDL No.: MFCD00002744

Synonymer:  Tetradecylic acid, Tetradecanoic acid
  • CAS nummer:544-63-8
  • Formel:C₁₄H₂₈O₂
  • MDL nummer:MFCD00002744
  • Molecular Weight:228.38 g/mol
  • Merck Index:14,6333
  • Kogende Pt:326 °C
  • Smeltende Pt:53…56 °C
  • Tæthed:0.862
  • Flash Pt:175 °C (347 °F)
  • Opbevaringstemperatur:Ambient
  • EINECS:208-875-2
Specifikationer

Specifikationer

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.