Order Entry
Denmark
Orders LinkContactUsLinkComponent
Hydroquinone ≥99%

Hydroquinone ≥99%

Glass bottle for solids, 5.000 g
Catalog # A11411.0I
Corrosive
Health
Irritant
Environment
Hydroquinone ≥99%
Catalog # A11411.0I
CAS Number:  123-31-9
Hydroquinone ≥99%
Hydroquinone ≥99%

Glass bottle for solids, 5.000 g

CAS Number:  123-31-9
Price
4.900,00 kr
Log in for availability
1 * 5.000 g
Product Details & Documents

Purity: 99% CAS No.: 123-31-9 Molecular Formula: C6H6O2 Formula Weight: 110.11 Flash Point: 165°(329°F) Odour: Odourless pH: 3.75 Melting Point: 170-172° Boiling Point: 285-287° Beilstein Reference: 605970 Merck Reference: 14,4808 UN No.: 3077 MDL No.: MFCD00002339

Synonyms:  1,4-Dihydroxybenzene, Quinol
  • CAS Number:123-31-9
  • Formula:C₆H₄(OH)₂
  • MDL Number:MFCD00002339
  • Molecular Weight:110.11 g/mol
  • Merck Index:14,4808
  • Boiling Pt:285…287 °C
  • Melting Pt:170…172 °C
  • Density:1.320
  • Flash Pt:165 °C (329 °F)
  • Storage Temperature:Ambient
  • EINECS:204-617-8
  • UN:3077
  • ADR:9,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5.000 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.