Order Entry
Denmark
Orders LinkContactUsLinkComponent
D-(-)-Serine ≥99%

D-(-)-Serine ≥99%

Glass bottle for solids, 100 g
Catalog # A11353.22
D-(-)-Serine ≥99%
Catalog # A11353.22
CAS Number:  312-84-5
D-(-)-Serine ≥99%
D-(-)-Serine ≥99%

Glass bottle for solids, 100 g

CAS Number:  312-84-5
Price
5.390,00 kr
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 99% CAS No.: 312-84-5 Molecular Formula: C3H7NO3 Formula Weight: 105.09 Melting Point: 218-220° dec. Beilstein Reference: 1721403 Merck Reference: 14,8460 Optical Rotation: -13° (c=5 in 5N HCl) MDL No.: MFCD00004269

Synonyms:  (R)-(-)-Serine, H-D-Ser-OH
  • CAS Number:312-84-5
  • Formula:C₃H₇NO₃
  • MDL Number:MFCD00004269
  • Molecular Weight:105.09 g/mol
  • Merck Index:14,8460
  • Melting Pt:218…220 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:206-229-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications