Order Entry
Denmark
Orders LinkContactUsLinkComponent
D-(-)-Serine ≥99%

D-(-)-Serine ≥99%

Glass bottle for solids, 5 g
Katalog # A11353.06
Leverandør:   Thermo Fisher Scientific
D-(-)-Serine ≥99%
Katalog # A11353.06
CAS nummer:  312-84-5
D-(-)-Serine ≥99%
D-(-)-Serine ≥99%

Glass bottle for solids, 5 g

CAS nummer:  312-84-5
Price
428,00 kr
Log in for availability
1 * 5 g
Product Details & Documents

Purity: 99% CAS No.: 312-84-5 Molecular Formula: C3H7NO3 Formula Weight: 105.09 Melting Point: 218-220° dec. Beilstein Reference: 1721403 Merck Reference: 14,8460 Optical Rotation: -13° (c=5 in 5N HCl) MDL No.: MFCD00004269

Synonyms:  (R)-(-)-Serine, H-D-Ser-OH
  • CAS Number:312-84-5
  • Formula:C₃H₇NO₃
  • MDL Number:MFCD00004269
  • Molecular Weight:105.09 g/mol
  • Merck Index:14,8460
  • Melting Pt:218…220 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:206-229-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.