Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Mesitylene ≥98%

Glass bottle, 100 mL
Catalog # A11323.AE
Flammable
Irritant
Environment
Mesitylene ≥98%
Catalog # A11323.AE
CAS Number:  108-67-8
Mesitylene ≥98%
Mesitylene ≥98%

Glass bottle, 100 mL

CAS Number:  108-67-8
Price
404,00 kr
Log in for availability
1 * 100 mL
Product Details & Documents

Purity: 98+% CAS No.: 108-67-8 Molecular Formula: C9H12 Formula Weight: 120.20 Flash Point: 44°(111°F) Refractive Index: 1.4990 Odour: Unpleasant Melting Point: -45° Boiling Point: 163-165° Beilstein Reference: 906806 Merck Reference: 14,5907 UN No.: 2325 MDL No.: MFCD00008538

Synonyms:  1,3,5-Trimethylbenzene
  • CAS Number:108-67-8
  • Formula:C₉H₁₂
  • MDL Number:MFCD00008538
  • Molecular Weight:120.19 g/mol
  • Merck Index:14,5907
  • Boiling Pt:163…165 °C
  • Melting Pt:–45 °C
  • Density:0.865
  • Flash Pt:44 °C (111 °F)
  • Storage Temperature:Ambient
  • EINECS:203-604-4
  • UN:2325
  • ADR:3,III
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Cat. No.:
    A11323.AE

Specifications