Order Entry
Denmark
Orders LinkContactUsLinkComponent
Paraformaldehyde ≥97%

Paraformaldehyde ≥97%

Glass bottle for solids, 10.000 g
Catalog # A11313.0C
Corrosive
Flammable
Health
Irritant
Paraformaldehyde ≥97%
Catalog # A11313.0C
CAS Number:  30525-89-4
Paraformaldehyde ≥97%
Paraformaldehyde ≥97%

Glass bottle for solids, 10.000 g

CAS Number:  30525-89-4
Price
3.530,00 kr
Log in for availability
1 * 10.000 g
Product Details & Documents
Synonyms:  Polyoxymethylene
  • CAS Number:30525-89-4
  • Formula:(CH₂O)n
  • MDL Number:MFCD00133991
  • Merck Index:14,7025
  • Boiling Pt:135…136 °C (13 mmHg)
  • Melting Pt:120…170 °C (dec.)
  • Density:1.30
  • Flash Pt:71 °C (159 °F)
  • Storage Temperature:Ambient
  • EINECS:200-001-8
  • UN:2213
  • ADR:4.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10.000 g

Specifications