Order Entry
Denmark
Orders LinkContactUsLinkComponent
1-Hexadecanol ≥98%

1-Hexadecanol ≥98%

Glass bottle for solids, 2.500 g
Catalog # A11180.0E
Irritant
1-Hexadecanol ≥98%
Catalog # A11180.0E
CAS Number:  36653-82-4
1-Hexadecanol ≥98%
1-Hexadecanol ≥98%

Glass bottle for solids, 2.500 g

CAS Number:  36653-82-4
Price
1.270,00 kr
Log in for availability
1 * 2.500 g
Product Details & Documents

Purity: 98% CAS No.: 36653-82-4 Molecular Formula: CH3(CH2)15OH Formula Weight: 242.45 Flash Point: 135°(275°F) Physical Form: Crystalline Density: 0.817 Melting Point: 47-51° Boiling Point: 190°/15mm Solubility: Soluble in alcohol, chloroform, ether Beilstein Reference: 1748475 Merck Reference: 14,2028 MDL No.: MFCD00004760

Synonyms:  Cetyl alcohol, Palmityl alcohol
  • CAS Number:36653-82-4
  • Formula:H₃C(CH₂)₁₄CH₂OH
  • MDL Number:MFCD00004760
  • Molecular Weight:242.45 g/mol
  • Merck Index:14,2028
  • Boiling Pt:190 °C (15 mmHg)
  • Melting Pt:47…51 °C
  • Density:0.817
  • Flash Pt:135 °C (275 °F)
  • Storage Temperature:Ambient
  • EINECS:253-149-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    2.500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.