Order Entry
Denmark
Orders LinkContactUsLinkComponent
Hydroxyurea ≥98%

Hydroxyurea ≥98%

Glass bottle for solids, 1 g
Katalog # A10831.03
Health
Leverandør:   Thermo Fisher Scientific
Hydroxyurea ≥98%
Katalog # A10831.03
CAS nummer:  127-07-1
Hydroxyurea ≥98%
Hydroxyurea ≥98%

Glass bottle for solids, 1 g

CAS nummer:  127-07-1
Price
376,00 kr
Log in for availability
1 * 1 g
Product Details & Documents

Purity: 98% CAS No.: 127-07-1 Molecular Formula: CH4N2O2 Formula Weight: 76.06 Melting Point: ca 140° dec. Beilstein Reference: 1741548 Merck Reference: 14,4848 MDL No.: MFCD00007943

An inhibitor of DNA synthesis and an anticancer agent

Synonyms:  N-(Aminocarbonyl)hydroxylamine, Hydroxycarbamide
  • CAS Number:127-07-1
  • Formula:CH₄N₂O₂
  • MDL Number:MFCD00007943
  • Molecular Weight:76.05 g/mol
  • Merck Index:14,4848
  • Boiling Pt:296 °C (1013 hPa)
  • Melting Pt:∼ 140 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:204-821-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications