Order Entry
Denmark
ContactUsLinkComponent
Flask volumetric, capacity 2l, class A, amber graduated, polystoppered, tolerance 0.6ml, height 370mm, NS-ground 29/32, graduation c olor white, conformity certified. 1 * 1 items
Catalog # HIRS264019437
Supplier:  HIRSCHMANN
Flask volumetric, capacity 2l, class A, amber graduated, polystoppered, tolerance 0.6ml, height 370mm, NS-ground 29/32, graduation c olor white, conformity certified. 1 * 1 items
Catalog # HIRS264019437
Supplier:  HIRSCHMANN
Frequently Bought Together