Flask volumetric, capacity 50ml, class A, amber graduated, polystoppered, tolerance 0.06ml, height 140mm, NS-ground 12/21, draduatio n color white, conformity certified. 1 * 1 items
Catalog # HIRS264017537
Supplier: HIRSCHMANN
Frequently Bought Together