Order Entry
Denmark
Orders LinkContactUsLinkComponent
1,2-Dichloroethane ≥99%

1,2-Dichloroethane ≥99%

Glass bottle, 500 mL
Catalog # A12775.AP
Flammable
Health
Irritant
1,2-Dichloroethane ≥99%
Catalog # A12775.AP
CAS Number:  107-06-2
1,2-Dichloroethane ≥99%
1,2-Dichloroethane ≥99%

Glass bottle, 500 mL

CAS Number:  107-06-2
Price
441,00 kr
Log in for availability
1 * 500 mL
Product Details & Documents

Purity: 99+% CAS No.: 107-06-2 Molecular Formula: C2H4Cl2 Formula Weight: 98.96 Flash Point: 15°(59°F) Refractive Index: 1.4448 Melting Point: -35° Boiling Point: 83° Beilstein Reference: 605264 Merck Reference: 14,3797 UN No.: 1184 MDL No.: MFCD00000963

Synonyms:  Ethylene chloride, Ethylene dichloride, Dichloroethane
  • CAS Number:107-06-2
  • Formula:ClH₂CCH₂Cl
  • MDL Number:MFCD00000963
  • Molecular Weight:98.96 g/mol
  • Merck Index:14,3797
  • Boiling Pt:83 °C
  • Melting Pt:–35 °C
  • Density:1.256
  • Flash Pt:15 °C (59 °F)
  • Storage Temperature:Ambient
  • EINECS:203-458-1
  • UN:1184
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL

Specifications