Order Entry
Denmark
Orders LinkContactUsLinkComponent
1,2-Dichloroethane ≥99%
Glass bottle, 2.500 mL
  A12775.0F
1,2-Dichloroethane ≥99%
  A12775.0F
 :  107-06-2
1,2-Dichloroethane ≥99%
1,2-Dichloroethane ≥99%
Glass bottle, 2.500 mL
 :  107-06-2
888,00 kr
1 * 2.500 mL
Purity: 99+% CAS No.: 107-06-2 Molecular Formula: C2H4Cl2 Formula Weight: 98.96 Flash Point: 15°(59°F) Refractive Index: 1.4448 Melting Point: -35° Boiling Point: 83° Beilstein Reference: 605264 Merck Reference: 14,3797 UN No.: 1184 MDL No.: MFCD00000963
 :  Ethylene chloride, Ethylene dichloride, Dichloroethane
  •  :107-06-2
  •  :ClH₂CCH₂Cl
  •  :MFCD00000963
  •  :98.96 g/mol
  •  :14,3797
  •  :83 °C
  •  :–35 °C
  •  :1.256
  •  :15 °C (59 °F)
  •  :Ambient
  •  :203-458-1
  •  :1184
  •  :3,II
  • Pack type:
    Glass bottle
  • Pk:
    2.500 mL
Anbefalinger