S-MONOVETTE AMMONIUM HEPARIN 4.5ML 1 * 500 items
SARS05.1105
: SARSTEDT
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Ethanol absolute ≥99.5% USP, Ph. Eur.
: VWR Chemicals
Ethanol 96% vol Ph. Eur.
: VWR Chemicals
VWR® Standard Line Transfer Pipettes
: VWR Collection
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® Serological Pipette, Standard Line
: VWR Collection
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection