Order Entry
Denmark
Orders LinkContactUsLinkComponent
Serological pipettes

Serological pipettes

25 ml, 0,2 ml, +, Red, 20/pack, 200/case
Catalog # SARS86.1685.020
Marketsource
Supplier:   SARSTEDT
Serological pipettes
Catalog # SARS86.1685.020
Supplier:  SARSTEDT
Marketsource
Serological pipettes

25 ml, 0,2 ml, +, Red, 20/pack, 200/case

Price
524,08 kr
Log in for availability
1 * 200 Items
Product Details & Documents

PS, graduated, sterile.

  • Plugged
  • Easy identification by colour code
  • Larger pipetting volume due to negative graduation
  • Optimised mouth piece for universal fit in most common pipettors
  • Easy to open, static-free packaging
Specifications
  • Colour code:
    Red
  • Packed:
    20/pack, 200/case
  • Sterile:
    +
  • Capacity:
    25 ml
  • Division:
    0,2 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.