Order Entry
Denmark
Orders LinkContactUsLinkComponent
Serological pipettes

Serological pipettes

1 ml, 0,01 ml, +, Yellow, Individually wrapped, 1000/case
Catalog # SARS86.1251.001
Marketsource
Supplier:   SARSTEDT
Serological pipettes
Catalog # SARS86.1251.001
Supplier:  SARSTEDT
Marketsource
Serological pipettes

1 ml, 0,01 ml, +, Yellow, Individually wrapped, 1000/case

Price
936,96 kr
Log in for availability
1 * 1.000 Items
Product Details & Documents

PS, graduated, sterile.

  • Plugged
  • Easy identification by colour code
  • Larger pipetting volume due to negative graduation
  • Optimised mouth piece for universal fit in most common pipettors
  • Easy to open, static-free packaging
Specifications
  • Colour code:
    Yellow
  • Packed:
    Individually wrapped, 1000/case
  • Sterile:
    +
  • Capacity:
    1 ml
  • Division:
    0,01 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.