Order Entry
Denmark
Orders LinkContactUsLinkComponent
Bootie cabinets

Bootie cabinets

Bootie Cabinet 56, 783×400×2100 mm
Catalog # 134-0478
Supplier:   Palbam
Bootie cabinets
Catalog # 134-0478
Supplier:  Palbam
Bootie cabinets
Bootie cabinets

Bootie Cabinet 56, 783×400×2100 mm

Price
Inquire For Price
Log in for availability
1 * 1 Items
Product Details & Documents

The modular bootie cabinets come in a variety of shelf lengths.

  • Supplied with leveler legs and foot covers
  • This symetric product can be used from either side
  • Adhesive labelling numbers supplied

Construction: Stainless steel 304L with electropolished finish

Delivery information: Typically available from stock for direct shipment from the supplier.

Specifications
  • Description:
    Bootie Cabinet 56
  • W×D×H:
    783×400×2100 mm